Gene Transcripts
- Gene Symbol
- DUSP5P1
- Entrez Gene ID
- 574029
- Gene Name
- dual specificity phosphatase 5 pseudogene 1
- Gene Aliases
- DUSP5P
- Chromosome Location
- Chr. 1: 228644910 - 228652412 on Build GRCh38
- UniGene ID
- Hs.647777
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002834.1 |
|
2 |
2180 |
|
GenBank mRNA |
AK092260.1 |
- |
2 |
2180 |
|
View Full Product Details