Gene Transcripts

Gene Symbol
TIPARP-AS1
Entrez Gene ID
100287227
Gene Name
TIPARP antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 3: 156673171 - 156675713 on Build GRCh38
UniGene ID
Hs.12813
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027954.1 1 2009
GenBank mRNA BC033324.1 - 1 1975
BC038770.1 - 1 643

View Full Product Details