Gene Transcripts
- Gene Symbol
- MTVR2
- Entrez Gene ID
- 246754
- Gene Name
- mouse mammary tumor virus receptor homolog 2
- Gene Aliases
- Chromosome Location
- Chr. 17: 56884102 - 56884913 on Build GRCh38
- UniGene ID
- Hs.720736
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027025.1 |
|
1 |
65 |
|
GenBank mRNA |
AA191116.1 |
- |
1 |
65 |
|
View Full Product Details