Gene Transcripts

Gene Symbol
CYP2B7P
Entrez Gene ID
1556
Gene Name
cytochrome P450 family 2 subfamily B member 7, pseudogene
Gene Aliases
CYP2B, CYP2B7, CYP2B7P1
Chromosome Location
Chr. 19: 40924265 - 40950660 on Build GRCh38
UniGene ID
Hs.529117
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001278.1 2, 3 343
GenBank mRNA AK307933.1 - 2, 3 389
AK310944.1 - 2, 3 393
DQ198366.1 - 2, 3 335
DQ198367.1 - 2, 3 335

View Full Product Details