Gene Transcripts
- Gene Symbol
- PIP5K1P1
- Entrez Gene ID
- 206426
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase type 1 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 6: 7986102 - 7990344 on Build GRCh38
- UniGene ID
- Hs.676966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027712.1 |
|
1 |
4110 |
|
GenBank mRNA |
BC028580.2 |
- |
1 |
4109 |
|
View Full Product Details