Gene Transcripts

Gene Symbol
PIP5K1P1
Entrez Gene ID
206426
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 pseudogene 1
Gene Aliases
Chromosome Location
Chr. 6: 7986102 - 7990344 on Build GRCh38
UniGene ID
Hs.676966
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027712.1 1 1944
GenBank mRNA AK312819.1 - 1 1549
BC028580.2 - 1 1943

View Full Product Details