Gene Transcripts
- Gene Symbol
- SLC6A10P
- Entrez Gene ID
- 386757
- Gene Name
- solute carrier family 6 member 10, pseudogene
- Gene Aliases
- CT-2, CT2, SLC6A10, SLC6A10pA
- Chromosome Location
- Chr. 16: 32877476 - 32885142 on Build GRCh38
- UniGene ID
- Hs.433146
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003083.2 |
|
9 |
3790 |
|
GenBank mRNA |
AK125843.1 |
- |
9 |
3791 |
|
- Gene Symbol
- SLC6A10PB
- Entrez Gene ID
- 653562
- Gene Name
- solute carrier family 6 member 8 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 16|NT_187383: 1139582 - 1147627 on Build GRCh38
- UniGene ID
- Hs.729445
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC068290.1 |
- |
4 |
757 |
|
View Full Product Details