Gene Transcripts
- Gene Symbol
- FAM92A1P2
- Entrez Gene ID
- 403315
- Gene Name
- family with sequence similarity 92, member A3
- Gene Aliases
- FAM92A3
- Chromosome Location
- Chr. 4: 183037665 - 183040119 on Build GRCh38
- UniGene ID
- Hs.652187
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI459302.1 |
- |
1 |
557 |
|
Chr. 4 - 183037665 - 183038350 [+] |
GenBankmRNA |
BI459384.1 |
- |
|
556 |
|
Chr. - - [-] |
View Full Product Details