Gene Transcripts

Gene Symbol
HHLA3
Entrez Gene ID
11147
Gene Name
HERV-H LTR-associating 3
Gene Aliases
Chromosome Location
Chr. 1: 70353840 - 70368023 on Build GRCh38
UniGene ID
Hs.142245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027404.1 2, 3 394
XM_011540548.2 XP_011538850.1 2, 3 293
GenBank mRNA AF126164.1 - 2, 3 375

View Full Product Details