Gene Transcripts
- Gene Symbol
- LOC100101478
- Entrez Gene ID
- 100101478
- Gene Name
- H2B histone family member M pseudogene
- Gene Aliases
- Chromosome Location
- Chr. X: 104060504 - 104062938 on Build GRCh38
- UniGene ID
- Hs.496530
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130172.1 |
|
3 |
609 |
|
GenBank mRNA |
AY730279.1 |
- |
3 |
609 |
|
BC028039.1 |
- |
4 |
440 |
|
- Gene Symbol
- H2BFXP
- Entrez Gene ID
- 767811
- Gene Name
- H2B histone family member X, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. X: 103975924 - 103978357 on Build GRCh38
- UniGene ID
- Hs.496530
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003238.2 |
|
3 |
609 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ217928.1 |
- |
3 |
628 |
|
Chr. X - 104060627 - 104062938 [-] |
View Full Product Details