Gene Transcripts

Gene Symbol
LINC01550
Entrez Gene ID
388011
Gene Name
long intergenic non-protein coding RNA 1550
Gene Aliases
C14orf64
Chromosome Location
Chr. 14: 97925610 - 97978124 on Build GRCh38
UniGene ID
Hs.651477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015430.1 2 377
GenBank mRNA AK091668.1 - 2 363
AK097943.1 - 2 232
BC043585.1 - 2 377
BX647564.1 - 2 361
JX073283.1 - 2 313


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DA521036.1 - 2 363 Chr. 14 - 97960263 - 97978110 [-]
GENCODE ENST00000512901.6 - 2 361 Chr. 14 - 97958036 - 97978108 [-]
GenBankmRNA BI772351.1 - 2 301 Chr. 14 - 97959933 - 97978046 [-]
GenBankmRNA BX109572.1 - 2 322 Chr. 14 - 97968300 - 97978070 [-]
GENCODE ENST00000556462.1 - 2 232 Chr. 14 - 97933062 - 97978056 [-]
GENCODE ENST00000499006.6 - 2 377 Chr. 14 - 97925610 - 97978124 [-]
GenBankmRNA AA418794.1 - 2 289 Chr. 14 - 97968315 - 97978037 [-]
GenBankmRNA DB154307.1 - 2 318 Chr. 14 - 97968842 - 97978065 [-]
GenBankmRNA BI834723.1 - 2 380 Chr. 14 - 97926352 - 97978124 [-]
GENCODE ENST00000502187.5 - 2 363 Chr. 14 - 97957930 - 97978110 [-]

View Full Product Details