Gene Transcripts
- Gene Symbol
- LINC01550
- Entrez Gene ID
- 388011
- Gene Name
- long intergenic non-protein coding RNA 1550
- Gene Aliases
- C14orf64
- Chromosome Location
- Chr. 14: 97925610 - 97978124 on Build GRCh38
- UniGene ID
- Hs.651477
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015430.1 |
|
3 |
608 |
|
GenBank mRNA |
BC043585.1 |
- |
3 |
608 |
|
JX073283.1 |
- |
3 |
544 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI834723.1 |
- |
3 |
611 |
|
Chr. 14 - 97926352 - 97978124 [-] |
View Full Product Details