Gene Transcripts
- Gene Symbol
- PCAT4
- Entrez Gene ID
- 118425
- Gene Name
- prostate cancer associated transcript 4 (non-protein coding)
- Gene Aliases
- GDEP, PCA4, PCAN1
- Chromosome Location
- Chr. 4: 79827471 - 79863247 on Build GRCh38
- UniGene ID
- Hs.104215
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026555.1 |
|
2 |
1716 |
|
GenBank mRNA |
AK056825.1 |
- |
2 |
1716 |
|
View Full Product Details