Gene Transcripts

Gene Symbol
KPNA1
Entrez Gene ID
3836
Gene Name
karyopherin subunit alpha 1
Gene Aliases
IPOA5, NPI-1, RCH2, SRP1
Chromosome Location
Chr. 3: 122421901 - 122514939 on Build GRCh38
UniGene ID
Hs.161008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026698.1 6 638
XM_005247439.3 XP_005247496.1 3 1145
XM_011512797.2 XP_011511099.1 6 2112
XM_017006363.1 XP_016861852.1 6 2086
XM_017006364.1 XP_016861853.1 6 515
GenBank mRNA AK125626.1 - 2 465
AK128280.1 - 3 1145
AK299030.1 - 5 524
AK312713.1 - 6 633
DN991078.1 - 6 480


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BI763343.1 - 6 592 Chr. 3 - 122457728 - 122514895 [-]
GenBankmRNA DC369049.1 - 5 524 Chr. 3 - 122457774 - 122496639 [-]
GenBankmRNA BX505713.1 - 2 233 Chr. 3 - 122451564 - 122459652 [-]
GenBankmRNA BQ434617.1 - 6 562 Chr. 3 - 122452003 - 122514863 [-]

View Full Product Details