Gene Transcripts
- Gene Symbol
- SPDYE8P
- Entrez Gene ID
- 728524
- Gene Name
- speedy/RINGO cell cycle regulator family member E8, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 7: 73019731 - 73029762 on Build GRCh38
- UniGene ID
- Hs.571275
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003664.2 |
|
1 |
41 |
|
GenBank mRNA |
BC045636.1 |
- |
1 |
44 |
|
BC068610.1 |
- |
1 |
44 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI562159.1 |
- |
1 |
49 |
|
Chr. 7 - 75307380 - 75309220 [+] |
View Full Product Details