Gene Transcripts
- Gene Symbol
- LINC01553
- Entrez Gene ID
- 283025
- Gene Name
- long intergenic non-protein coding RNA 1553
- Gene Aliases
- C10orf40
- Chromosome Location
- Chr. 10: 59958217 - 59960913 on Build GRCh38
- UniGene ID
- Hs.522955
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024340.1 |
|
2 |
484 |
|
GenBank mRNA |
BC038741.1 |
- |
2 |
484 |
|
BC048976.2 |
- |
1 |
422 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG723969.1 |
- |
2 |
489 |
|
Chr. 10 - 59959479 - 59960913 [-] |
GenBankmRNA |
DB026117.1 |
- |
2 |
481 |
|
Chr. 10 - 59959463 - 59960910 [-] |
GenBankmRNA |
BI463999.1 |
- |
2 |
489 |
|
Chr. 10 - 59959182 - 59960913 [-] |
GenBankmRNA |
DB444108.2 |
- |
2 |
484 |
|
Chr. 10 - 59959375 - 59960913 [-] |
GenBankmRNA |
DB023414.1 |
- |
2 |
481 |
|
Chr. 10 - 59959492 - 59960910 [-] |
View Full Product Details