Gene Transcripts

Gene Symbol
C21orf62-AS1
Entrez Gene ID
54067
Gene Name
C21orf62 antisense RNA 1
Gene Aliases
C21orf49
Chromosome Location
Chr. 21: 32772100 - 32797707 on Build GRCh38
UniGene ID
Hs.54725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024622.1 4 852
NR_024623.1 4 840
GenBank mRNA AK096601.1 - 4 841
BC034794.1 - 2 566

View Full Product Details