Gene Transcripts

Gene Symbol
RHPN1-AS1
Entrez Gene ID
78998
Gene Name
RHPN1 antisense RNA 1 (head to head)
Gene Aliases
C8orf51
Chromosome Location
Chr. 8: 143366623 - 143368635 on Build GRCh38
UniGene ID
Hs.245886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026785.1 1 537
GenBank mRNA AK026503.1 - 1 448
BC000203.1 - 1 450
BI829214.1 - 1 215

View Full Product Details