Gene Transcripts
- Gene Symbol
- FAM86JP
- Entrez Gene ID
- 100125556
- Gene Name
- family with sequence similarity 86, member A pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 3: 125916601 - 125930024 on Build GRCh38
- UniGene ID
- Hs.553949
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG178286.1 |
- |
3 |
344 |
|
Chr. 3 - 125916628 - 125929031 [+] |
GenBankmRNA |
AL530538.3 |
- |
3 |
365 |
|
Chr. 3 - 125916625 - 125929195 [+] |
View Full Product Details