Gene Transcripts

Gene Symbol
FAM86JP
Entrez Gene ID
100125556
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
Chromosome Location
Chr. 3: 125916601 - 125930024 on Build GRCh38
UniGene ID
Hs.553949
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024250.1 4 357
NR_024251.1 4 455
GenBank mRNA BC032772.1 - 4 343
BC041080.1 - 4 426
BM781991.1 - 4 357

View Full Product Details