Gene Transcripts
- Gene Symbol
- LINC00483
- Entrez Gene ID
- 55018
- Gene Name
- long intergenic non-protein coding RNA 483
- Gene Aliases
- C17orf73
- Chromosome Location
- Chr. 17: 50761034 - 50767515 on Build GRCh38
- UniGene ID
- Hs.389460
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK000670.1 |
- |
5 |
941 |
|
View Full Product Details