Gene Transcripts
- Gene Symbol
- SKINT1L
- Entrez Gene ID
- 391037
- Gene Name
- Skint1 like (pseudogene)
- Gene Aliases
- BTN12, SKINT1, SKINTL, SKINTP
- Chromosome Location
- Chr. 1: 48101715 - 48182428 on Build GRCh38
- UniGene ID
- Hs.585393
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026749.2 |
|
4 |
361 |
|
GenBank mRNA |
DN994130.1 |
- |
4 |
369 |
|
View Full Product Details