Gene Transcripts
- Gene Symbol
- GUSBP3
- Entrez Gene ID
- 653188
- Gene Name
- glucuronidase, beta pseudogene 3
- Gene Aliases
- Chromosome Location
- Chr. 5: 69639463 - 69710527 on Build GRCh38
- UniGene ID
- Hs.631974
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027386.2 |
|
6 |
931 |
|
- Gene Symbol
- SMA4
- Entrez Gene ID
- 11039
- Gene Name
- glucuronidase beta pseudogene
- Gene Aliases
- SMA3, b55C20.2
- Chromosome Location
- Chr. 5: 70127462 - 70290177 on Build GRCh38
- UniGene ID
- Hs.582500
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X83300.1 |
- |
4 |
672 |
|
View Full Product Details