Gene Transcripts
- Gene Symbol
- SNORA7B
- Entrez Gene ID
- 677797
- Gene Name
- small nucleolar RNA, H/ACA box 7B
- Gene Aliases
- ACA7B
- Chromosome Location
- Chr. 3: 129397210 - 129397348 on Build GRCh38
- UniGene ID
- Hs.711399
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002992.2 |
|
1 |
112 |
|
View Full Product Details