Gene Transcripts
- Gene Symbol
- SNORA50A
- Entrez Gene ID
- 677830
- Gene Name
- small nucleolar RNA, H/ACA box 50A
- Gene Aliases
- ACA50, SNORA50, SNORA76A
- Chromosome Location
- Chr. 16: 58559796 - 58559931 on Build GRCh38
- UniGene ID
- Hs.677515
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002980.1 |
|
1 |
110 |
|
GenBank mRNA |
AJ609468.1 |
- |
1 |
110 |
|
View Full Product Details