Gene Transcripts
- Gene Symbol
- GSTTP2
- Entrez Gene ID
- 653399
- Gene Name
- glutathione S-transferase theta pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.652398
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003082.1 |
|
5 |
2025 |
|
GenBank mRNA |
AK127991.1 |
- |
5 |
2024 |
|
View Full Product Details