Gene Transcripts
- Gene Symbol
- CYP1B1-AS1
- Entrez Gene ID
- 285154
- Gene Name
- CYP1B1 antisense RNA 1
- Gene Aliases
- C2orf58
- Chromosome Location
- Chr. 2: 38131105 - 38181855 on Build GRCh38
- UniGene ID
- Hs.353894
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027252.1 |
|
3 |
679 |
|
GenBank mRNA |
BC031410.1 |
- |
3 |
679 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI909894.1 |
- |
3 |
672 |
|
Chr. 2 - 38131112 - 38180958 [+] |
GenBankmRNA |
BI754396.1 |
- |
3 |
679 |
|
Chr. 2 - 38131107 - 38180918 [+] |
View Full Product Details