Gene Transcripts

Gene Symbol
CYP1B1-AS1
Entrez Gene ID
285154
Gene Name
CYP1B1 antisense RNA 1
Gene Aliases
C2orf58
Chromosome Location
Chr. 2: 38131105 - 38181855 on Build GRCh38
UniGene ID
Hs.353894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027252.1 3 780
GenBank mRNA BC031410.1 - 3 780

View Full Product Details