Gene Transcripts
- Gene Symbol
- CYP1B1-AS1
- Entrez Gene ID
- 285154
- Gene Name
- CYP1B1 antisense RNA 1
- Gene Aliases
- C2orf58
- Chromosome Location
- Chr. 2: 38131105 - 38181855 on Build GRCh38
- UniGene ID
- Hs.353894
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027252.1 |
|
3 |
1052 |
|
GenBank mRNA |
BC031410.1 |
- |
3 |
1052 |
|
View Full Product Details