Gene Transcripts
- Gene Symbol
- PCDHGB8P
- Entrez Gene ID
- 56120
- Gene Name
- protocadherin gamma subfamily B, 8 pseudogene
- Gene Aliases
- PCDH-PSI3
- Chromosome Location
- Chr. 5: 141426286 - 141428258 on Build GRCh38
- UniGene ID
- Hs.672506
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001297.1 |
|
1 |
978 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF152529.3 |
- |
1 |
978 |
|
Chr. 5 - 141426286 - 141428258 [+] |
View Full Product Details