Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|---|
RefSeq | NR_002824.3 | 34 | 5160 | |||
GenBank mRNA | AB002391.2 | - | 31 | 4847 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
34 | 5166 | ||
AF041081.1 | - | LIT2191 HERC2P3 |
30 | 5123 | ||
AK095014.1 | - | 10 | 2288 | |||
CR749409.1 | - | 23 | 3602 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_036443.1 | 12 | 1785 | ||
GenBank mRNA | AL832775.1 | - | 1 | 3863 | |
BC047911.1 | - | 12 | 1785 |