Gene Transcripts

Gene Symbol
PPY2P
Entrez Gene ID
23614
Gene Name
pancreatic polypeptide 2, pseudogene
Gene Aliases
PPY2
Chromosome Location
Chr. 17: 28247444 - 28248289 on Build GRCh38
UniGene ID
Hs.744962
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002181.1 1 141
GenBank mRNA AF085945.1 - 1 36
AF222903.1 - 1 141


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX280431.1 - 1 36 Chr. 17 - 28247549 - 28248216 [+]
GenBankmRNA CR747376.1 - 1 36 Chr. 17 - 28247549 - 28248210 [+]

View Full Product Details