Gene Transcripts
- Gene Symbol
- RBMY3AP
- Entrez Gene ID
- 64593
- Gene Name
- RNA binding motif protein, Y-linked, family 3, member A pseudogene
- Gene Aliases
- RBMY3P, RBMY4P, SPATA14
- Chromosome Location
- Chr. Y: 9610721 - 9615153 on Build GRCh38
- UniGene ID
- Hs.404172
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001573.2 |
|
3 |
274 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
U94386.3 |
- |
3 |
274 |
|
Chr. Y - 9610721 - 9615153 [-] |
View Full Product Details