Gene Transcripts
- Gene Symbol
- BCAR4
- Entrez Gene ID
- 400500
- Gene Name
- breast cancer anti-estrogen resistance 4 (non-protein coding)
- Gene Aliases
- Chromosome Location
- Chr. 16: 11819830 - 11828832 on Build GRCh38
- UniGene ID
- Hs.24611
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000574028.1 |
- |
1 |
64 |
|
Chr. 16 - 11819948 - 11828818 [-] |
GenBankmRNA |
R80133.1 |
- |
|
51 |
|
Chr. - - [-] |
GenBankmRNA |
R67448.1 |
- |
|
74 |
|
Chr. - - [-] |
GENCODE |
ENST00000571259.5 |
- |
1 |
74 |
|
Chr. 16 - 11819829 - 11828828 [-] |
GenBankmRNA |
R67456.1 |
- |
|
61 |
|
Chr. - - [-] |
View Full Product Details