Gene Transcripts
- Gene Symbol
- SNAP47
- Entrez Gene ID
- 116841
- Gene Name
- synaptosome associated protein 47
- Gene Aliases
- C1orf142, ESFI5812, HEL-S-290, HEL170, SNAP-47, SVAP1
- Chromosome Location
- Chr. 1: 227728168 - 227781231 on Build GRCh38
- UniGene ID
- Hs.325081
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA176792.1 |
- |
2 |
548 |
|
Chr. 1 - 227728619 - 227732787 [+] |
GenBankmRNA |
DA046891.1 |
- |
3 |
683 |
|
Chr. 1 - 227728647 - 227732821 [+] |
GenBankmRNA |
DA053725.1 |
- |
2 |
481 |
|
Chr. 1 - 227728686 - 227732843 [+] |
View Full Product Details