Gene Transcripts
- Gene Symbol
- SCARNA14
- Entrez Gene ID
- 692149
- Gene Name
- small Cajal body-specific RNA 14
- Gene Aliases
- U100
- Chromosome Location
- Chr. 15: 66347206 - 66347342 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_004388.1 |
|
1 |
41 |
|
View Full Product Details