Gene Transcripts
- Gene Symbol
- PA2G4P4
- Entrez Gene ID
- 647033
- Gene Name
- proliferation-associated 2G4 pseudogene 4
- Gene Aliases
- PA2G4L1
- Chromosome Location
- Chr. 3: 156809271 - 156812021 on Build GRCh38
- UniGene ID
- Hs.745109
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003284.1 |
|
1 |
1941 |
|
GenBank mRNA |
AK025367.1 |
- |
1 |
1948 |
|
View Full Product Details