Gene Transcripts
- Gene Symbol
- NEAT1
- Entrez Gene ID
- 283131
- Gene Name
- nuclear paraspeckle assembly transcript 1 (non-protein coding)
- Gene Aliases
- LINC00084, NCRNA00084, TncRNA, VINC
- Chromosome Location
- Chr. 11: 65422798 - 65445540 on Build GRCh38
- UniGene ID
- Hs.523789
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK026198.1 |
- |
1 |
91 |
|
Chr. 11 - 65422804 - 65423809 [+] |
View Full Product Details