Gene Transcripts
- Gene Symbol
- Vis1
- Entrez Gene ID
- 110153
- Gene Name
- viral integration site 1
- Gene Aliases
- Gm27276, His-1, His1
- Chromosome Location
- Chr. 2: 45238980 - 45282900 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U10269.1 |
- |
LIT1767 His1 |
2, 3 |
306 |
|
View Full Product Details