Gene Transcripts
A Gene Symbol is not available because the non-coding siRNA was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID. Non-coding siRNAs are designed to RefSeq (NR_) or to curated sequences from RNAdb represented by GenBank RNA or ESTdb accessions.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG920590.1 |
- |
4 |
601 |
|
Chr. X - 103606955 - 103615193 [-] |
View Full Product Details