Gene Transcripts

Gene Symbol
RBMY3AP
Entrez Gene ID
64593
Gene Name
RNA binding motif protein, Y-linked, family 3, member A pseudogene
Gene Aliases
RBMY3P, RBMY4P, SPATA14
Chromosome Location
Chr. Y: 9610721 - 9615153 on Build GRCh38
UniGene ID
Hs.404172
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA U94385.1 - LIT2106
RBMIII pseudogene
2, 3 164

View Full Product Details