Gene Transcripts
- Gene Symbol
- STYXL1
- Entrez Gene ID
- 51657
- Gene Name
- serine/threonine/tyrosine interacting-like 1
- Gene Aliases
- DUSP24, MK-STYX, MKSTYX
- Chromosome Location
- Chr. 7: 75995820 - 76048709 on Build GRCh38
- UniGene ID
- Hs.11615
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
RNADB ID |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY927581.1 |
- |
LIT3580 AY927581 |
8 |
805 |
|
View Full Product Details