Gene Transcripts
- Gene Symbol
- LINC01194
- Entrez Gene ID
- 404663
- Gene Name
- long intergenic non-protein coding RNA 1194
- Gene Aliases
- CT49, TAG
- Chromosome Location
- Chr. 5: 12574857 - 12805183 on Build GRCh38
- UniGene ID
- Hs.552273
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY328030.1 |
- |
3 |
1142 |
|
View Full Product Details