Gene Transcripts

Gene Symbol
KCNN3
Entrez Gene ID
3782
Gene Name
potassium calcium-activated channel subfamily N member 3
Gene Aliases
KCa2.3, SK3, SKCA3, hSK3
Chromosome Location
Chr. 1: 154697462 - 154870278 on Build GRCh38
UniGene ID
Hs.490765
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204087.1 NP_001191016.1 9 7716
NM_002249.5 NP_002240.3 8 7671
NM_170782.2 NP_740752.1 8 6592
GenBank mRNA BC030100.1 - 1 416

View Full Product Details