Gene Transcripts
- Gene Symbol
- DUSP5P1
- Entrez Gene ID
- 574029
- Gene Name
- dual specificity phosphatase 5 pseudogene 1
- Gene Aliases
- DUSP5P
- Chromosome Location
- Chr. 1: 228644910 - 228652412 on Build GRCh38
- UniGene ID
- Hs.647777
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055963.1 |
- |
1 |
2211 |
|
- Gene Symbol
- LOC105373132
- Entrez Gene ID
- 105373132
- Gene Name
- uncharacterized LOC105373132
- Gene Aliases
- Chromosome Location
- Chr. 1: 228646284 - 228650007 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details