Gene Transcripts

Gene Symbol
TTLL11
Entrez Gene ID
158135
Gene Name
tubulin tyrosine ligase like 11
Gene Aliases
C9orf20, bA244O19.1
Chromosome Location
Chr. 9: 121815674 - 122093606 on Build GRCh38
UniGene ID
Hs.438937
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005251728.2 XP_005251785.1 8 6534
XM_005251729.4 XP_005251786.1 9 7611
GenBank mRNA AK123875.1 - 1 544

View Full Product Details