Gene Transcripts
- Gene Symbol
- ZNF518A
- Entrez Gene ID
- 9849
- Gene Name
- zinc finger protein 518A
- Gene Aliases
- ZNF518
- Chromosome Location
- Chr. 10: 96129220 - 96165512 on Build GRCh38
- UniGene ID
- Hs.600823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095317.1 |
- |
1 |
2830 |
|
- Gene Symbol
- TDRP
- Entrez Gene ID
- 157695
- Gene Name
- testis development related protein
- Gene Aliases
- C8orf42, Inm01, TDRP1, TDRP2
- Chromosome Location
- Chr. 8: 489790 - 545781 on Build GRCh38
- UniGene ID
- Hs.289293
- Species
- Homo sapiens
View Full Product Details