Gene Transcripts
- Gene Symbol
- TTC39B
- Entrez Gene ID
- 158219
- Gene Name
- tetratricopeptide repeat domain 39B
- Gene Aliases
- C9orf52
- Chromosome Location
- Chr. 9: 15167204 - 15307360 on Build GRCh38
- UniGene ID
- Hs.563630
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC070139.1 |
- |
4 |
400 |
|
View Full Product Details