Gene Transcripts

Gene Symbol
ZNF91
Entrez Gene ID
7644
Gene Name
zinc finger protein 91
Gene Aliases
HPF7, HTF10
Chromosome Location
Chr. 19: 23305004 - 23395560 on Build GRCh38
UniGene ID
Hs.531632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090762.1 - 1 1937

View Full Product Details