Gene Transcripts

Gene Symbol
ORMDL1
Entrez Gene ID
94101
Gene Name
ORMDL sphingolipid biosynthesis regulator 1
Gene Aliases
Chromosome Location
Chr. 2: 189766149 - 189784371 on Build GRCh38
UniGene ID
Hs.700632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011512200.2 XP_011510502.1 3 2233
GenBank mRNA AK126336.1 - 1 1436

View Full Product Details